missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Grace™ Vydac™ 218TP C18 218TP HPLC Columns
Vydac™ TP HPLC Columns are the industry's standard wide pore media range for peptide and protein separations. The 300 Å TP silica gives polypeptide molecules complete access to the interior of the silica pores.
POA
Specifications
Certifications/Compliance | ISO 9001, 14000 and 13485 procedure, FDA 21 CFR 820 for medical devices |
---|---|
Film Thickness | 5 μm |
Hydrophobicity | Yes |
Length (Metric) | 100 mm |
Particle Shape | Spherical |
Product Code | Brand | Column Format | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Column Format | Diameter (Metric) | Price | Quantity & Availability | ||||
11369990
|
Grace™
218TP51010 |
Preparative Column | 10 mm |
POA
Each |
N/A | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11358890
|
Grace™
218TP5410 |
Analytical Column | 4.6 mm |
POA
Each |
N/A | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
Description
- Long column lifetime and negligible phase leaching
- Reliable protein purifications, scalable from analytical to preparative scale
- Referenced in a large number of patents and publications
Specifications
ISO 9001, 14000 and 13485 procedure, FDA 21 CFR 820 for medical devices | |
Yes | |
Spherical | |
300 Å | |
L1 | |
Silica | |
Reversed Phase | |
Reversed Phase |
5 μm | |
100 mm | |
5 μm | |
60 m2/g | |
C18 | |
8% | |
Yes | |
Silica |