missing translation for 'onlineSavingsMsg'
Learn More

Grace™ Vydac™ 218TP C18 218TP HPLC Columns

Vydac™ TP HPLC Columns are the industry's standard wide pore media range for peptide and protein separations. The 300 Å TP silica gives polypeptide molecules complete access to the interior of the silica pores.

Brand:  Grace™ 218TP51010

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11369990

  • POA / Each
Estimated Shipment: 19-06-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
Specifications

Specifications

ISO 9001, 14000 and 13485 procedure, FDA 21 CFR 820 for medical devices
Preparative Column
5 μm
100 mm
5 μm
0.6 mL/g
L1
Silica
Reversed Phase
Reversed Phase
Preparative Column
Octadecyl Silane
10 mm
Yes
Spherical
300 Å
60 m2/g
C18
8%
Yes
Silica
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions