missing translation for 'onlineSavingsMsg'
Learn More

Cabozantinib (S)-malate, Thermo Scientific™

Product Code. 17505565 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 17505565

Brand: Thermo Scientific Acros 468900010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1140909-48-3
Molecular Formula C32H30FN3O10
Molecular Weight (g/mol) 635.60
MDL Number MFCD20923480
InChI Key HFCFMRYTXDINDK-UHFFFAOYNA-N
IUPAC Name 2-hydroxybutanedioic acid; N'1-{4-[(6,7-dimethoxyquinolin-4-yl)oxy]phenyl}-N1-(4-fluorophenyl)cyclopropane-1,1-dicarboxamide
SMILES OC(CC(O)=O)C(O)=O.COC1=C(OC)C=C2C(OC3=CC=C(NC(=O)C4(CC4)C(=O)NC4=CC=C(F)C=C4)C=C3)=CC=NC2=C1

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Crystalline Powder
Color White to Light Beige
Melting Point 166°C to 172°C
Quantity 1 g
Chemical Name or Material Cabozantinib (S)-malate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.