Learn More
ASTM D2622/D7039, Low Sulfur Curve, VHG™
Click to view available options
Quantity:
6 x 100 mL
Unit Size:
600mL
Description
ASTM D2622/D7039, Low Sulfur Curve
Specifications
Specifications
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | 20 cSt Mineral Oil |
| Quantity | 6 x 100 mL |