missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Standard: Mg @ 1000 μg/g in Hydrocarbon Oil, VHG™
Click to view available options
Quantity:
50 g
200 g
Unit Size:
200g
50g
Description
Magnesium Standard: Mg @ 1000 μg/g in Hydrocarbon Oil
Specifications
Specifications
| Chemical Name or Material | Magnesium |
| CAS | 7439-95-4 |
| Methods | ICP/XRF/RDE |
| Analyte or Component Names | Magnesium |
| Shipping Condition | Room Temperature |
| Molecular Formula | Mg |
| Certifications/Compliance | ISO 17034 |
| Synonym | Mg |
| InChI Formula | InChI=1S/Mg |
| SMILES | [Mg] |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction