missing translation for 'onlineSavingsMsg'
Learn More

1-(Mesitylenesulfonyl)imidazole, 98+%, Thermo Scientific™

Product Code. 11457327
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11457327

Brand: Thermo Scientific Alfa Aesar A15053.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50257-39-1
Molecular Formula C12H14N2O2S
Molecular Weight (g/mol) 250.32
MDL Number MFCD00005284
InChI Key XFHVQSPNDDOABS-UHFFFAOYSA-N
Synonym 1-mesitylsulfonyl-1h-imidazole, 1-2-mesitylenesulfonyl imidazole, n-mesitylenesulfonyl imidazole, n-mesitylenesulfonylimidazole, 1-2,4,6-trimethylbenzenesulfonyl imidazole, 1-2,4,6-trimethylphenyl sulphonyl-1h-imidazole, 1h-imidazole, 1-2,4,6-trimethylphenyl sulfonyl, 2,4,6-trimethylphenyl sulfonyl imidazole, acmc-209kkw, mesitylenesulfonylimidazole
PubChem CID 99556
IUPAC Name 1-(2,4,6-trimethylphenyl)sulfonylimidazole
SMILES CC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=CN=C1

Specifications

Melting Point 97°C to 99°C
Quantity 100 g
Sensitivity Moisture sensitive
Formula Weight 250.32
Percent Purity ≥98%
Chemical Name or Material 1-(Mesitylenesulfonyl)imidazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.