accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 50257-39-1

CH3CH3H3CSOONN

CAS RN 50257-39-1

IUPAC Name: 1-(2,4,6-trimethylbenzenesulfonyl)-1H-imidazole
Synonyms: 1-(2,4,6-trimethylbenzenesulfonyl)imidazole
Molecular Weight (g/mol): 250.32
Molecular Formula: C12H14N2O2S
InChi Key: XFHVQSPNDDOABS-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=CN=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results