accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 97964-56-2

H3CNCH3OHOONHOClCl

CAS RN 97964-56-2

IUPAC Name: 4-[(3,4-dichlorophenyl)formamido]-4-(dipentylcarbamoyl)butanoic acid
Synonyms: lorglumide 氯戊米特
Molecular Weight (g/mol): 459.41
Molecular Formula: C22H32Cl2N2O4
InChi Key: IEKOTSCYBBDIJC-UHFFFAOYNA-N
SMILES: CCCCCN(CCCCC)C(=O)C(CCC(O)=O)NC(=O)C1=CC=C(Cl)C(Cl)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results