missing translation for 'onlineSavingsMsg'
Learn More

Lorglumide sodium salt, 98+%, Thermo Scientific™

Product Code. 15494249
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 15494249

Brand: Thermo Scientific Alfa Aesar J61713.MB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent and specific non-peptide cholecystokinin (CCK) receptor antagonist

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97964-56-2
Molecular Formula C22H32Cl2N2O4
Molecular Weight (g/mol) 459.41
MDL Number MFCD00083183
InChI Key IEKOTSCYBBDIJC-UHFFFAOYNA-N
Synonym lorglumide, lorglumide inn, lorglumidum latin, lorglumida spanish, ccris 6857, chembl24938, d,l-4-3,4-dichlorobenzoylamino-5-dipentylamino-5-oxo-pentanoic acid, lorglumide sodium salt, lorglumida, lorglumidum
PubChem CID 3960
IUPAC Name 4-[(3,4-dichlorobenzoyl)amino]-5-(dipentylamino)-5-oxopentanoic acid
SMILES CCCCCN(CCCCC)C(=O)C(CCC(O)=O)NC(=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Color White
Quantity 25 mg
Formula Weight 481.39
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Lorglumide sodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.