CAS RN 96-48-0
CAS RN 96-48-0
IUPAC Name:
oxolan-2-one
Synonyms:
4-butyrolactone butyrylactone 4-deoxytetronate gamma-butyryllactone gamma butyrolactone BLO g-hydroxybutyric acid lactone 4-butanolide lactone, 4-hydroxybutyric acid 1,4-lactone
Molecular Weight (g/mol):
86.09
Molecular Formula:
C4H6O2
InChi Key:
YEJRWHAVMIAJKC-UHFFFAOYSA-N
SMILES:
O=C1CCCO1
1
–
2
of
2
results
1
Giemsa-Romanowski Solution, Penta Chemicals
Top quality chemicals for laboratories, schools, hospitals, research institutes, and large manufacturing plants.
2
Gammabutyrolactone (GBL), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.