Learn More
Gammabutyrolactone (GBL), Mikromol™
Description
Gammabutyrolactone (GBL)
Specifications
Specifications
| Chemical Name or Material | gamma-Butyrolactone (GBL) |
| CAS | 96-48-0 |
| Analyte or Component Names | gamma-Butyrolactone (GBL) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C4 H6 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | gamma-Butyrolactone, GBL, Gammabutyrolactone |
| InChI Formula | InChI=1S/C4H6O2/c5-4-2-1-3-6-4/h1-3H2 |
| SMILES | O=C1CCCO1 |
| IUPAC Name | oxolan-2-one |
| Show More |