CAS RN 95959-20-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
∆9,11-Dehydro-17beta-estradiol 17-Valerate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Hydroxyestra-1,3,5(10),9(11)-tetraen-17beta-yl Pentanoate (9,11-Didehydroestradiol Valerate), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
∆9,11-Dehydro-17beta-estradiol 17-Valerate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
∆9,11-Dehydro-17beta-estradiol 17-Valerate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.