Learn More
∆9,11-Dehydro-17beta-estradiol 17-Valerate, TRC
Specifications
Specifications
| Chemical Name or Material | 9,11-Didehydroestradiol Valerate |
| CAS | 95959-20-9 |
| Molecular Formula | C23 H30 O3 |
| Quantity | 2.5 mg |
| Synonym | 3-Hydroxyestra-1,3,5(10),9(11)-tetraen-17beta-yl Pentanoate, Estradiol Valerate Imp. C (EP), 9,11-Didehydroestradiol Valerate, Estradiol-9-ene valerate |
| InChI Formula | InChI=1S/C23H30O3/c1-3-4-5-22(25)26-21-11-10-20-19-8-6-15-14-16(24)7-9-17(15)18(19)12-13-23(20,21)2/h7,9,12,14,19-21,24H,3-6,8,10-11,13H2,1-2H3/t19-,20+,21+,23+/m1/s1 |
| SMILES | CCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCc4cc(O)ccc4C3=CC[C@]12C |
| IUPAC Name | [(8S,13S,14S,17S)-3-hydroxy-13-methyl-6,7,8,12,14,15,16,17-octahydrocyclopenta[a]phenanthren-17-yl] pentanoate |
| Molecular Weight (g/mol) | 354.48 |
| Formula Weight | 354.2195 |