accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BNOHOOH

CAS RN 929250-35-1

IUPAC Name: [6-(benzyloxy)pyridin-3-yl]boronic acid
Synonyms: 6-(benzyloxy)pyridin-3-ylboronic acid
Molecular Weight (g/mol): 229.04
Molecular Formula: C12H12BNO3
InChi Key: CUZOGAUUSKBDAO-UHFFFAOYSA-N
SMILES: OB(O)C1=CN=C(OCC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)