missing translation for 'onlineSavingsMsg'
Learn More

6-Benzyloxypyridine-3-boronic acid, 97%, Thermo Scientific™

Product Code. 11966471 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11966471 5 g 5g
11956471 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11966471 Supplier Thermo Scientific Acros Supplier No. 445320050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 929250-35-1
Molecular Formula C12H12BNO3
Molecular Weight (g/mol) 229.04
MDL Number MFCD06798262
InChI Key CUZOGAUUSKBDAO-UHFFFAOYSA-N
Synonym 2-benzyloxypyridine-5-boronic acid, 6-benzyloxypyridine-3-boronic acid, 6-benzyloxy pyridin-3-yl boronic acid, 6-benzyloxy pyridine-3-boronic acid, 6-benzyloxy pyridin-3-ylboronic acid, 2-benzyloxy-pyridine-5-boronic acid, 6-benzyloxy pyridin-3-yl-3-boronic acid, 2-benzyloxypyridin-5-yl boronic acid, 2-benzyloxy pyridine-5-ylboronic acid
PubChem CID 17750216
IUPAC Name (6-phenylmethoxypyridin-3-yl)boronic acid
SMILES OB(O)C1=CN=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 107°C to 110°C
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 229.04
Percent Purity 97%
Chemical Name or Material 6-Benzyloxypyridine-3-boronic acid
compliance-icons
Product Identifier
  • 6-Benzyloxypyridine-3-boronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.