accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 871-83-0

H3CCH3CH3

CAS RN 871-83-0

IUPAC Name: 2-methylnonane
Synonyms: 2-methylnonane C9-11 isoparaffin C10-13 isoparaffin
Molecular Weight (g/mol): 142.29
Molecular Formula: C10H22
InChi Key: SGVYKUFIHHTIFL-UHFFFAOYSA-N
SMILES: CCCCCCCC(C)C
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results