CAS RN 855300-09-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-Acetylphenyl Ethyl(methyl)carbamate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Des [3-(1-Dimethylamino)ethyl] 3-Acetyl Rivastigmine, TRC
CAS: 855300-09-3 Molecular Formula: C12 H15 N O3 Molecular Weight (g/mol): 221.25 Synonym: 3-Acetylphenyl Ethyl(methyl)carbamate,Des-[3-(1-Dimethylamino)ethyl] 3-Acetylrivastigmine,Rivastigmine Related Compound D (USP),Rivastigmine Tartrate Imp. C (EP) IUPAC Name: (3-acetylphenyl) N-ethyl-N-methylcarbamate SMILES: CCN(C)C(=O)Oc1cccc(c1)C(=O)C