Learn More
Des [3-(1-Dimethylamino)ethyl] 3-Acetyl Rivastigmine, TRC
Product Code. 30358389
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358389 100 mg 100mg
30354299 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30358389 Supplier Toronto Research Chemicals Supplier No. TRCD289320100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 855300-09-3
Molecular Formula C12 H15 N O3
Molecular Weight (g/mol) 221.25
Synonym 3-Acetylphenyl Ethyl(methyl)carbamate, Des-[3-(1-Dimethylamino)ethyl] 3-Acetylrivastigmine, Rivastigmine Related Compound D (USP), Rivastigmine Tartrate Imp. C (EP)
IUPAC Name (3-acetylphenyl) N-ethyl-N-methylcarbamate
SMILES CCN(C)C(=O)Oc1cccc(c1)C(=O)C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C12H15NO3/c1-4-13(3)12(15)16-11-7-5-6-10(8-11)9(2)14/h5-8H,4H2,1-3H3
Formula Weight 221.1052
Chemical Name or Material 3-Acetylphenyl Ethyl(methyl)carbamate