CAS RN 845273-93-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sevelamer Carbonate, Technical Grade, TRC
CAS: 845273-93-0 Molecular Formula: C3 H7 N . C3 H5 Cl O . C H2 O3 Molecular Weight (g/mol): 211.64 Synonym: Renvela,Genz 644470,GT 335-012,Carbonic Acid, Compound With (Chloromethyl)oxirane Polymer With 2-Propen-1-amine IUPAC Name: carbonic acid;2-(chloromethyl)oxirane;prop-2-en-1-amine SMILES: NCC=C.OC(=O)O.ClCC1CO1