Learn More
Sevelamer Carbonate, Technical Grade, TRC
Product Code. 17924325
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17924325 100 mg 100mg
17902845 500 mg 500mg
17922545 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17924325 Supplier Toronto Research Chemicals Supplier No. TRCS039370100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 845273-93-0
Molecular Formula C3 H7 N . C3 H5 Cl O . C H2 O3
Molecular Weight (g/mol) 211.64
Synonym Renvela, Genz 644470, GT 335-012, Carbonic Acid, Compound With (Chloromethyl)oxirane Polymer With 2-Propen-1-amine
IUPAC Name carbonic acid;2-(chloromethyl)oxirane;prop-2-en-1-amine
SMILES NCC=C.OC(=O)O.ClCC1CO1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C3H5ClO.C3H7N.CH2O3/c4-1-3-2-5-3;1-2-3-4;2-1(3)4/h3H,1-2H2;2H,1,3-4H2;(H2,2,3,4)
Formula Weight 211.0611
Chemical Name or Material Sevelamer Carbonate