CAS RN 70954-04-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(1-Deoxy-D-fructos-1-yl)-L-threonine, TRC
CAS: 70954-04-0 Molecular Formula: C10H19NO8 Molecular Weight (g/mol): 281.26 Synonym: [R-(R*,S*)]-1-[(1-Carboxy-2-hydroxypropyl)amino]-1-deoxy-D-fructose IUPAC Name: (((3S,4R,5R)-2,3,4,5-tetrahydroxytetrahydro-2H-pyran-2-yl)methyl)-L-threonine and(((3S,4S,5R)-2,3,4-trihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)methyl)-L-threonine SMILES: OC1(CN[C@H](C(O)=O)[C@H](O)C)[C@@H](O)[C@H](O)[C@H](O)CO1→O=C(O)[C@@H](NCC2(O)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)C