Learn More
N-(1-Deoxy-D-fructos-1-yl)-L-threonine, TRC
Product Code. 30365089
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365089 50 mg 50mg
30360597 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30365089 Supplier Toronto Research Chemicals Supplier No. TRCD23591050MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 70954-04-0
Molecular Formula C10H19NO8
Molecular Weight (g/mol) 281.26
Synonym [R-(R*,S*)]-1-[(1-Carboxy-2-hydroxypropyl)amino]-1-deoxy-D-fructose
IUPAC Name (((3S,4R,5R)-2,3,4,5-tetrahydroxytetrahydro-2H-pyran-2-yl)methyl)-L-threonine and(((3S,4S,5R)-2,3,4-trihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)methyl)-L-threonine
SMILES OC1(CN[C@H](C(O)=O)[C@H](O)C)[C@@H](O)[C@H](O)[C@H](O)CO1→O=C(O)[C@@H](NCC2(O)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)C
Specifications
Quantity 50 mg
InChI Formula InChI=1S/2C10H19NO8/c1-4(12)6(9(16)17)11-3-10(18)8(15)7(14)5(13)2-19-10;1-4(13)6(9(16)17)11-3-10(18)8(15)7(14)5(2-12)19-10/h2*4-8,11-15,18H,2-3H2,1H3,(H,16,17)/t4-,5+,6-,7+,8+,10?;4-,5-,6-,7-,8+,10?/m11/s1
Formula Weight 281.11
Percent Purity >95%
Chemical Name or Material N-(1-Deoxy-D-fructos-1-yl)-L-threonine
Grade HPLC