CAS RN 690646-00-5
CAS RN 690646-00-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(3-Chlorophenylsulfonylamino)cyclohexanecarboxylic acid, 96%, Thermo Scientific™
CAS: 690646-00-5 Molecular Formula: C13H16ClNO4S MDL Number: MFCD20040116 Synonym: 1-3-chlorophenylsulfonamido cyclohexanecarboxylic acid,1-3-chlorophenyl sulfonamido cyclohexane-1-carboxylic acid,1-3-chlorophenyl sulfonylamino cyclohexane-1-carboxylic acid,1-3-chlorobenzenesulfonamido cyclohexane-1-carboxylic acid