CAS RN 66-02-4
CAS RN 66-02-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,5-Diiodo-DL-tyrosine, TRC
CAS: 66-02-4 Molecular Formula: C9 H9 I2 N O3 Molecular Weight (g/mol): 432.98 Synonym: Tyrosine, 3,5-diiodo-,3,5-Diiodotyrosine,3,5-Diiodo-4-hydroxyphenylalanine,Agontan,Apothyrin,Cemiod,DIT,DL-3,5-Diiodotyrosine,Dityrin,Flaianina,Gorgoic acid, diiodo-,Gorgonic acid, diiodo-,Iodogorgoic acid,Iodogorgonic acid,Itir,Jodgorgon,NSC 208959,NSC 97936,β-(4-Hydroxy-3,5-diiodophenyl)alanine,3,5-Diiodo-DL-tyrosine,(2RS)-2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid IUPAC Name: 2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid SMILES: NC(Cc1cc(I)c(O)c(I)c1)C(=O)O