Learn More
3,5-Diiodo-DL-tyrosine, TRC
Product Code. 30385592
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385592 100 mg
30372191 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30385592 Supplier Toronto Research Chemicals Supplier No. TRCD455175100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 66-02-4
Molecular Formula C9 H9 I2 N O3
Molecular Weight (g/mol) 432.98
Synonym Tyrosine, 3,5-diiodo-, 3,5-Diiodotyrosine, 3,5-Diiodo-4-hydroxyphenylalanine, Agontan, Apothyrin, Cemiod, DIT, DL-3,5-Diiodotyrosine, Dityrin, Flaianina, Gorgoic acid, diiodo-, Gorgonic acid, diiodo-, Iodogorgoic acid, Iodogorgonic acid, Itir, Jodgorgon, NSC 208959, NSC 97936, β-(4-Hydroxy-3,5-diiodophenyl)alanine, 3,5-Diiodo-DL-tyrosine, (2RS)-2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid
IUPAC Name 2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid
SMILES NC(Cc1cc(I)c(O)c(I)c1)C(=O)O
Specifications
InChI Formula InChI=1S/C9H9I2NO3/c10-5-1-4(2-6(11)8(5)13)3-7(12)9(14)15/h1-2,7,13H,3,12H2,(H,14,15)
Quantity 100 mg
Formula Weight 432.8672
Shipping Condition Room Temperature
Chemical Name or Material 3,5-Diiodo-DL-tyrosine