CAS RN 656820-32-5
CAS RN 656820-32-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
STEMCELL Technologies Reversine
Adenosine receptor, non-muscle myosin II (NM II), MEK1, and aurora kinase inhibitor
2
Reversine, MedChemExpress
MedChemExpress Reversine is a novel class of ATP-competitive Aurora kinase inhibitor with IC50s of 400, 500 and 400 nM for Aurora A, Aurora B and Aurora C, respectively.