Learn More
Reversine, MedChemExpress
Product Code. 16492267
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16492267
Brand: Medchem Express HY14711
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Reversine is a novel class of ATP-competitive Aurora kinase inhibitor with IC50s of 400, 500 and 400 nM for Aurora A, Aurora B and Aurora C, respectively.
Reversine is a novel class of ATP-competitive Aurora kinase inhibitor with IC50s of 400, 500 and 400 nM for Aurora A, Aurora B and Aurora C, respectively.
Specifications
Chemical Name or Material Reversine
Physical Form Solid
CAS 656820-32-5
Color Light Green
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C21H27N7O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 20 mg/mL (50.83 mM; Need ultrasonic)
SMILES C1(NC2=CC=C(N3CCOCC3)C=C2)=NC(NC4CCCCC4)=C5C(NC=N5)=N1
Molecular Weight (g/mol) 393.49
Formula Weight 393.49
Percent Purity 99.22%
Grade Research