Learn More
STEMCELL Technologies Reversine
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
- Reversine is a 2,6-disubstituted purine derivative that inhibits the human A3 adenosine receptor, nonmuscle myosin II heavy chain, mitogen activated extracellular signal regulated kinase-1 (MEK1), and Aurora B kinase (Chen et al.; Perreira et al.; D'Alise et al.)
Specifications
Specifications
| CAS | 656820-32-5 |
| Quantity | 5 mg |
| Molecular Formula | C21H27N7O |
| Purity | >98% |