accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 65646-68-6

H3CH3CH3CONHHOH3CCH3(E)(E)(E)(E)

CAS RN 65646-68-6

IUPAC Name: (2E,4E,6E,8E)-N-(4-hydroxyphenyl)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenamide
Synonyms: fenretinide 13-cis-fenretinide 4 hydroxyphenylretinamide fenretinidum 13 cis isomer fenretinide 4-hydroxyphenylretinamide 4HPR HPR fenretinida 4-hydroxy(phenyl)retinamide
Molecular Weight (g/mol): 391.56
Molecular Formula: C26H33NO2
InChi Key: AKJHMTWEGVYYSE-FXILSDISSA-N
SMILES: C\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(=O)NC1=CC=C(O)C=C1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results