Learn More
Fenretinide, MedChemExpress
Product Code. 16466746
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466746 100 mg 100mg
16452577 50 mg 50mg
16426478 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16466746 Supplier Medchem Express Supplier No. HY15373
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
MedChemExpress Fenretinide (4-HPR) is a synthetic retinoid deriverative, binding to the retinoic acid receptors (RAR) at concentrations necessary to induce cell death.
Fenretinide (4-HPR) is a synthetic retinoid deriverative, binding to the retinoic acid receptors (RAR) at concentrations necessary to induce cell death.
Specifications
Chemical Name or Material Fenretinide
CAS 65646-68-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H33NO2
Quantity 100 mg
Synonym 4-HPR
Solubility Information DMSO : ≥ 130 mg/mL (332.01 mM)
SMILES CC(/C=C/C=C(/C=C/C1=C(CCCC(C)1C)C)C)=C\C(NC2=CC=C(O)C=C2)=O
Molecular Weight (g/mol) 391.55
Formula Weight 391.55
Percent Purity 99.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death