CAS RN 63199-74-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-(tert-Butylamino)propiophenone Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Deschloro Bupropion Hydrochloride, TRC
CAS: 63199-74-6 Molecular Formula: C13 H19 N O . Cl H Molecular Weight (g/mol): 241.76 Synonym: 2-(tert-Butylamino)propiophenone Hydrochloride IUPAC Name: 2-(tert-butylamino)-1-phenylpropan-1-one;hydrochloride SMILES: Cl.CC(NC(C)(C)C)C(=O)c1ccccc1