Learn More
Deschloro Bupropion Hydrochloride, TRC
Product Code. 30368022
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368022 100 mg 100mg
30358383 50 mg 50mg
30366314 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30368022 Supplier Toronto Research Chemicals Supplier No. TRCD288545100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 63199-74-6
Molecular Formula C13 H19 N O . Cl H
Molecular Weight (g/mol) 241.76
Synonym 2-(tert-Butylamino)propiophenone Hydrochloride
IUPAC Name 2-(tert-butylamino)-1-phenylpropan-1-one;hydrochloride
SMILES Cl.CC(NC(C)(C)C)C(=O)c1ccccc1
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C13H19NO.ClH/c1-10(14-13(2,3)4)12(15)11-8-6-5-7-9-11;/h5-10,14H,1-4H3;1H
Formula Weight 241.1233
Percent Purity >95%
Chemical Name or Material 2-(tert-Butylamino)propiophenone Hydrochloride
Grade HPLC