accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 623-53-0

H3COOOCH3

CAS RN 623-53-0

IUPAC Name: ethyl methyl carbonate
Synonyms: ethyl methyl carbonate
Molecular Weight (g/mol): 104.11
Molecular Formula: C4H8O3
InChi Key: JBTWLSYIZRCDFO-UHFFFAOYSA-N
SMILES: CCOC(=O)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results