accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 62232-46-6

ClH3CH2NO

CAS RN 62232-46-6

IUPAC Name: [4-(2-benzylphenoxy)butyl](methyl)azanium chloride
Synonyms: bifemelane
Molecular Weight (g/mol): 305.85
Molecular Formula: C18H24ClNO
InChi Key: MEAHDXWXNNDSAK-UHFFFAOYSA-N
SMILES: [Cl-].C[NH2+]CCCCOC1=CC=CC=C1CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results