missing translation for 'onlineSavingsMsg'
Learn More

Bifemelane hydrochloride, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18732654
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18732654 10 mg 10mg
18722654 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18732654 Supplier Tocris Bioscience Supplier No. 0767/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MAO-A and MAO-B inhibitor

Bifemelane hydrochloride is an antidepressant MAO inhibitor. Reverses catalepsy induced by Tetrabenazine (Cat. No. 2175) in mice and increases locomotor activity in MPTP-treated marmosets.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62232-46-6
Molecular Formula C18H24ClNO
Molecular Weight (g/mol) 305.846
InChI Key MEAHDXWXNNDSAK-UHFFFAOYSA-N
Synonym bifemelane hydrochloride, celeport, alnert, unii-2n52s01ufy, 4-2-benzylphenoxy-n-methylbutan-1-amine hydrochloride, n-methyl-4-2-phenylmethyl phenoxy-1-butanamine hydrochloride, regid_for_cid_6917789, 4-2-benzylphenoxy-n-methylbutylamine hydrochloride
PubChem CID 6917789
IUPAC Name 4-(2-benzylphenoxy)-N-methylbutan-1-amine;hydrochloride
SMILES CNCCCCOC1=CC=CC=C1CC2=CC=CC=C2.Cl

Specifications

Quantity 10 mg
Formula Weight 305.84
Percent Purity >99%
Chemical Name or Material Bifemelane hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.