CAS RN 621-71-6
CAS RN 621-71-6
IUPAC Name:
1,3-bis(decanoyloxy)propan-2-yl decanoate
Synonyms:
tricaprin tridecanoin tri- (8CI) decanoin glyceryl tricaprate capric acid triglyceride decanoin, tri- decanoin, tri- (8CI) glycerol tricaprate dynasan 110 1,2,3-propanol tridecanoate
Molecular Weight (g/mol):
554.85
Molecular Formula:
C33H62O6
InChi Key:
LADGBHLMCUINGV-UHFFFAOYSA-N
SMILES:
CCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCC)OC(=O)CCCCCCCCC
1
–
9
of
9
results
2
Tricaprin 8000 μg/mL in Pyridine, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
5
Tricaprin 8000 μg/mL in Pyridine, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality