accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 62-13-5

HClH3CNHOOHHO

CAS RN 62-13-5

IUPAC Name: hydrogen 1-(3,4-dihydroxyphenyl)-2-(methylamino)ethan-1-one chloride
Synonyms: adrenone hydrochloride
Molecular Weight (g/mol): 217.65
Molecular Formula: C9H12ClNO3
InChi Key: CSRRBDMYOUQTCO-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CNCC(=O)C1=CC=C(O)C(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results