CAS RN 6170-42-9
CAS RN 6170-42-9
IUPAC Name:
hydrogen N-[(4-chlorophenyl)methyl]-N-[2-(dimethylamino)ethyl]pyridin-2-amine chloride
Synonyms:
hydrogen chloropyramine chloride
Molecular Weight (g/mol):
326.27
Molecular Formula:
C16H21Cl2N3
InChi Key:
VEYWWAGBHABATA-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CN(C)CCN(CC1=CC=C(Cl)C=C1)C1=CC=CC=N1
1
–
4
of
4
results
2
Chloropyramine Hydrochloride, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
3
Chloropyramine Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4
Chloropyramine hydrochloride, MedChemExpress
MedChemExpress Chloropyramine hydrochloride is a histamine receptor H1 antagonist which can also inhibit the biochemical function of VEGFR-3 and FAK.