Learn More
Chloropyramine Hydrochloride, TRC
Product Code. 30012556
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012556 1 g 1g
30008951 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30012556 Supplier Toronto Research Chemicals Supplier No. TRCC3812681G
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6170-42-9
Molecular Formula C16 H20 Cl N3 . Cl H
Molecular Weight (g/mol) 326.26
Synonym Chloropyramine Hydrochloride, 1,2-Ethanediamine, N1-[(4-chlorophenyl)methyl]-N2,N2-dimethyl-N1-2-pyridinyl-, hydrochloride (1:1), 1,2-Ethanediamine, N-[(4-chlorophenyl)methyl]-N',N'-dimethyl-N-2-pyridinyl-, monohydrochloride (9CI), Pyridine, 2-[(p-chlorobenzyl)[2-(dimethylamino)ethyl]amino]-, monohydrochloride (8CI), Alergosan, Allergan S hydrochloride, Chloropyramine hydrochloride, Chloropyribenzamine hydrochloride, Halopyramine hydrochloride, N-(β-Dimethylaminoethyl)-N-(p-chlorobenzyl)-α-aminopyridine hydrochloride, NSC 409949, Sinopen hydrochloride, Suprastin hydrochloride
IUPAC Name N'-[(4-chlorophenyl)methyl]-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine;hydrochloride
SMILES Cl.CN(C)CCN(Cc1ccc(Cl)cc1)c2ccccn2
Specifications
Quantity 1 g
InChI Formula InChI=1S/C16H20ClN3.ClH/c1-19(2)11-12-20(16-5-3-4-10-18-16)13-14-6-8-15(17)9-7-14;/h3-10H,11-13H2,1-2H3;1H
Formula Weight 325.1113
Percent Purity >95%
Chemical Name or Material Chloropyramine Hydrochloride
Grade HPLC