CAS RN 60397-77-5
CAS RN 60397-77-5
Form-2',4'-xylidide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-(2,4-Dimethylphenyl)formamide, TRC
CAS: 60397-77-5 Molecular Formula: C9 H11 N O Molecular Weight (g/mol): 149.19 Synonym: Formamide, N-(2,4-dimethylphenyl)-,N-(2,4-Dimethylphenyl)formamide,2,4-Dimethylformanilide,2,4-Dimethylphenylformamide,BTS 27919,N-Formyl-2,4-dimethylaniline,NSC 406488,o,p-Dimethylformanilide,Form-2',4'-xylidide IUPAC Name: N-(2,4-dimethylphenyl)formamide SMILES: Cc1ccc(NC=O)c(C)c1
N-(2,4-Dimethylphenyl)formamide 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality