Learn More
N-(2,4-Dimethylphenyl)formamide, TRC
Product Code. 18032320
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18032320 1 g 1g
18000740 100 mg 100mg
17959709 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18032320 Supplier Toronto Research Chemicals Supplier No. TRCD4767201G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 60397-77-5
Molecular Formula C9 H11 N O
Molecular Weight (g/mol) 149.19
Synonym Formamide, N-(2,4-dimethylphenyl)-, N-(2,4-Dimethylphenyl)formamide, 2,4-Dimethylformanilide, 2,4-Dimethylphenylformamide, BTS 27919, N-Formyl-2,4-dimethylaniline, NSC 406488, o,p-Dimethylformanilide, Form-2',4'-xylidide
IUPAC Name N-(2,4-dimethylphenyl)formamide
SMILES Cc1ccc(NC=O)c(C)c1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C9H11NO/c1-7-3-4-9(10-6-11)8(2)5-7/h3-6H,1-2H3,(H,10,11)
Formula Weight 149.0841
Chemical Name or Material Form-2',4'-Xylidide