CAS RN 546-18-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cholanoic Acid, TRC
CAS: 546-18-9 Molecular Formula: C24H40O2 Molecular Weight (g/mol): 360.57 Synonym: 5beta-Cholan-24-oic acid,5beta-Cholanic acid,5beta-Cholanoic acid,(5beta,17beta)-gamma-Methyl-androstane-17-butanoic Acid,L 596205-0,NSC 18161,Ursocholanic Acid IUPAC Name: (4R)-4-[(5S,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid SMILES: C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4CCCC[C@]4(C)[C@H]3CC[C@]12C