Learn More
Cholanoic Acid, TRC
Product Code. 30361225
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361225 10 mg 10mg
30355977 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30361225 Supplier Toronto Research Chemicals Supplier No. TRCC43143010MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 546-18-9
Molecular Formula C24H40O2
Molecular Weight (g/mol) 360.57
Synonym 5beta-Cholan-24-oic acid, 5beta-Cholanic acid, 5beta-Cholanoic acid, (5beta,17beta)-gamma-Methyl-androstane-17-butanoic Acid, L 596205-0, NSC 18161, Ursocholanic Acid
IUPAC Name (4R)-4-[(5S,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4CCCC[C@]4(C)[C@H]3CC[C@]12C
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C24H40O2/c1-16(7-12-22(25)26)19-10-11-20-18-9-8-17-6-4-5-14-23(17,2)21(18)13-15-24(19,20)3/h16-21H,4-15H2,1-3H3,(H,25,26)/t16-,17+,18+,19-,20+,21+,23+,24-/m1/s1
Formula Weight 360.3028
Chemical Name or Material Cholanoic Acid