accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 523-31-9

OOOO

CAS RN 523-31-9

IUPAC Name: 1,2-dibenzyl benzene-1,2-dicarboxylate
Synonyms: dibenzyl phthalate
Molecular Weight (g/mol): 346.38
Molecular Formula: C22H18O4
InChi Key: UCVPKAZCQPRWAY-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)C1=CC=CC=C1C(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results