CAS RN 50739-76-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-Amino-3,5-dibromo-benzenemethanol, TRC
CAS: 50739-76-9 Molecular Formula: C7 H7 Br2 N O Molecular Weight (g/mol): 280.94 Synonym: (2-Amino-3,5-dibromophenyl)methanol,Bromhexine Hydrochloride Imp. A (EP),Ambroxol Hydrochloride Imp. A (EP) IUPAC Name: (2-amino-3,5-dibromophenyl)methanol SMILES: Nc1c(Br)cc(Br)cc1CO
2
(2-Amino-3,5-dibromophenyl)methanol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.