Learn More
(2-Amino-3,5-dibromophenyl)methanol, Mikromol™
Description
(2-Amino-3,5-dibromophenyl)methanol
Specifications
Specifications
| Chemical Name or Material | (2-Amino-3,5-dibromophenyl)methanol |
| CAS | 50739-76-9 |
| Analyte or Component Names | (2-Amino-3,5-dibromophenyl)methanol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H7 Br2 N O |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Amino-3,5-dibromobenzyl Alcohol, Ambroxol Hydrochloride Imp. A (EP), Ambroxol Imp. A (EP), Bromhexine Hydrochloride Imp. A (EP), Bromhexine Imp. A (EP), (2-Amino-3,5-dibromophenyl)methanol, Ambroxol Hydrochloride Impurity A, Bromhexine Hydrochloride Impurity A, Ambroxol Impurity A, Bromhexine Impurity A |
| InChI Formula | InChI=1S/C7H7Br2NO/c8-5-1-4(3-11)7(10)6(9)2-5/h1-2,11H,3,10H2 |
| SMILES | Nc1c(Br)cc(Br)cc1CO |
| IUPAC Name | (2-amino-3,5-dibromophenyl)methanol |
| Show More |