CAS RN 484-78-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Hydroxy-DL-Kynurenine, TRC
CAS: 484-78-6 Molecular Formula: C10 H12 N2 O4 Molecular Weight (g/mol): 224.21 Synonym: α,2-Diamino-3-hydroxy-γ-oxo-benzenebutanoic acid,3-(3-Hydroxyanthraniloyl)-alanine,3-Hydroxykynurenine IUPAC Name: 2-amino-4-(2-amino-3-hydroxy-phenyl)-4-oxo-butanoic acid SMILES: NC(CC(=O)c1cccc(O)c1N)C(=O)O