Learn More
3-Hydroxy-DL-Kynurenine, TRC
Product Code. 30006600
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006600 10 mg 10mg
30004633 5 mg 5mg
30003863 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006600 Supplier Toronto Research Chemicals Supplier No. TRCH95075810MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 484-78-6
Molecular Formula C10 H12 N2 O4
Molecular Weight (g/mol) 224.21
Synonym α,2-Diamino-3-hydroxy-γ-oxo-benzenebutanoic acid, 3-(3-Hydroxyanthraniloyl)-alanine, 3-Hydroxykynurenine
IUPAC Name 2-amino-4-(2-amino-3-hydroxy-phenyl)-4-oxo-butanoic acid
SMILES NC(CC(=O)c1cccc(O)c1N)C(=O)O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C10H12N2O4/c11-6(10(15)16)4-8(14)5-2-1-3-7(13)9(5)12/h1-3,6,13H,4,11-12H2,(H,15,16)
Formula Weight 224.0797
Percent Purity >95%
Chemical Name or Material 3-Hydroxy-DL-kynurenine
Grade HPLC