accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COONN

CAS RN 38922-77-9

IUPAC Name: ethyl imidazo[1,2-a]pyridine-2-carboxylate
Synonyms: ethyl imidazo[1,2-a]pyridine-2-carboxylate
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: GNFACXDTRBVZJE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results