missing translation for 'onlineSavingsMsg'
Learn More

Ethyl imidazo[1,2-a]pyridine-2-carboxylate, 95%, Thermo Scientific™

Product Code. 15417517
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417517 10 g 10g
15407517 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417517 Supplier Thermo Scientific Alfa Aesar Supplier No. H35384.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38922-77-9
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.20
MDL Number MFCD02325394
InChI Key GNFACXDTRBVZJE-UHFFFAOYSA-N
Synonym ethyl imidazo 1,2-a pyridine-2-carboxylate, imidazo 1,2-a pyridine-2-carboxylic acid ethyl ester, ethyl h-imidazo 1,2-a pyridine-2-carboxylate, imidazo 1,2-a pyridine-2-carboxylic acid, ethyl ester, imidazo 1,2-a pyridine-2-carboxylicacid, ethyl ester, ethyl 4-hydroimidazo 1,2-a pyridine-2-carboxylate, pubchem17885, acmc-209j2i, ethyl imidazo 1,2-a picolinate, ethylimidazo 1,2-a pyridine-2-carboxylate
PubChem CID 818807
IUPAC Name ethyl imidazo[1,2-a]pyridine-2-carboxylate
SMILES CCOC(=O)C1=CN2C=CC=CC2=N1

Specifications

Melting Point 78°C to 80°C
Quantity 10 g
Formula Weight 190.2
Percent Purity 95%
Chemical Name or Material Ethyl imidazo[1,2-a]pyridine-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.